Avantor® Hichrom HI-624 MS, GC Columns
Supplier: VWR International
76431-276 76431-278 76431-280 76431-282 76431-284 76431-286 76431-288 76431-290 76433-352 76433-354 76433-356 76433-358 76433-360 76433-362 76433-364 76433-366 76433-368 76433-370 76433-372 76433-374 76433-376 76433-378 76433-380 76433-382 76433-384 76433-386 76433-388 76433-390 76433-392 76433-394 76433-396 76433-398 76433-400 76433-402 76433-404 76433-406 76433-408 76433-410 76433-412 76433-414 76433-416 76433-418 76433-420 76433-422 76433-424 76433-426 76433-428 76433-430 76433-432 76433-434 76433-436 76433-438 76433-440 76433-442 76433-444 76433-446 76433-448 76433-450 76433-452 76433-454 76433-456 76433-458 76433-460 76433-462 76433-464 76433-466 76433-468 76433-470 76433-472 76433-474 76433-476 76433-478 76433-480 76433-482 76433-484 76433-486 76433-488 76433-490 76433-492 76433-494 76433-496 76433-498 76433-500 76433-502 76433-504 76433-506 76433-508 76433-510 76433-512 76433-514 76433-516 76433-518 76433-520 76433-522 76433-524 76433-526 76433-528 76433-530 76433-532 76433-534 76433-536 76433-538 76433-540 76433-542 76433-544 76433-546 76433-548 76433-550 76433-552 76433-554 76433-556 76433-558 76433-560 76433-562 76433-564 76433-566 76433-568 76433-570 76433-572 76433-574 76433-576 76433-578 76433-580 76433-582 76433-584 76433-586 76433-588 76433-590 76433-592 76434-078 76434-080 76434-082 76434-084 76434-086 76434-088 76434-090 76434-092 76434-094 76434-096 76434-098 76434-100 76434-102 76434-104 76434-106 76434-108 76434-110 76434-112 76434-114 76434-116 76434-118
76431-276EA 1109.45 USD
76431-276 76431-278 76431-280 76431-282 76431-284 76431-286 76431-288 76431-290 76433-352 76433-354 76433-356 76433-358 76433-360 76433-362 76433-364 76433-366 76433-368 76433-370 76433-372 76433-374 76433-376 76433-378 76433-380 76433-382 76433-384 76433-386 76433-388 76433-390 76433-392 76433-394 76433-396 76433-398 76433-400 76433-402 76433-404 76433-406 76433-408 76433-410 76433-412 76433-414 76433-416 76433-418 76433-420 76433-422 76433-424 76433-426 76433-428 76433-430 76433-432 76433-434 76433-436 76433-438 76433-440 76433-442 76433-444 76433-446 76433-448 76433-450 76433-452 76433-454 76433-456 76433-458 76433-460 76433-462 76433-464 76433-466 76433-468 76433-470 76433-472 76433-474 76433-476 76433-478 76433-480 76433-482 76433-484 76433-486 76433-488 76433-490 76433-492 76433-494 76433-496 76433-498 76433-500 76433-502 76433-504 76433-506 76433-508 76433-510 76433-512 76433-514 76433-516 76433-518 76433-520 76433-522 76433-524 76433-526 76433-528 76433-530 76433-532 76433-534 76433-536 76433-538 76433-540 76433-542 76433-544 76433-546 76433-548 76433-550 76433-552 76433-554 76433-556 76433-558 76433-560 76433-562 76433-564 76433-566 76433-568 76433-570 76433-572 76433-574 76433-576 76433-578 76433-580 76433-582 76433-584 76433-586 76433-588 76433-590 76433-592 76434-078 76434-080 76434-082 76434-084 76434-086 76434-088 76434-090 76434-092 76434-094 76434-096 76434-098 76434-100 76434-102 76434-104 76434-106 76434-108 76434-110 76434-112 76434-114 76434-116 76434-118
Avantor® Hichrom HI-624 MS, GC Columns
Chromatography Columns
General purpose low-to-mid-polarity phase for a wide range of applications, including (for example): Residual solvents, volatile pollutants. HI-624 MS is an MS grade crossbonded column.
  • Phase composition: Crossbonded 6% cyanopropylphenyl, 94% methyl polysiloxane - MS grade
  • Long lifetime
Max temperature limits may change depending on the film thickness.