Avantor® Hichrom HI-5 MS Plus, GC Columns
Supplier: VWR International
76431-212 76431-214 76431-216 76431-218 76431-220 76431-222 76431-224 76431-226 76431-228 76431-230 76431-232 76431-234 76432-876 76432-878 76432-880 76432-882 76432-884 76432-886 76432-888 76432-890 76432-892 76432-894 76432-896 76432-898 76432-900 76432-902 76432-904 76432-906 76432-908 76432-910 76432-912 76432-914 76432-916 76432-918 76432-920 76432-922 76432-924 76432-926 76432-928 76432-930 76432-932 76432-934 76432-936 76432-938 76432-940 76432-942 76432-944 76432-946 76432-948 76432-950 76432-952 76432-954 76432-956 76432-958 76432-960 76432-962 76432-964 76432-966 76432-968 76432-970 76432-972 76432-974 76432-976 76432-978 76432-980 76432-982 76432-984 76432-986 76432-988 76432-990 76432-992 76432-994 76432-996 76432-998 76433-000 76433-002 76433-004 76433-006 76433-008 76433-010 76433-012 76433-014 76433-016 76433-018 76433-020 76433-022 76433-024 76433-026 76433-028 76433-030 76433-032 76433-034 76433-036 76433-038 76433-040 76433-042 76433-044 76433-046 76433-048 76433-050 76433-052 76433-054 76433-056 76433-058 76433-060 76433-062 76433-064 76433-066 76433-068 76433-070 76433-072 76433-074 76433-076 76433-078 76433-080 76433-082 76433-084 76433-086 76433-088 76433-090 76433-092 76433-094 76433-096 76433-098 76433-100 76433-102 76433-104 76433-106 76433-108 76433-110 76433-112 76434-010 76434-012 76434-014 76434-016 76434-018 76434-020 76434-022 76434-024 76434-026 76434-028 76434-030 76434-032 76434-034 76434-036 76434-038 76434-040 76434-042
76431-212EA 1577.38 USD
76431-212 76431-214 76431-216 76431-218 76431-220 76431-222 76431-224 76431-226 76431-228 76431-230 76431-232 76431-234 76432-876 76432-878 76432-880 76432-882 76432-884 76432-886 76432-888 76432-890 76432-892 76432-894 76432-896 76432-898 76432-900 76432-902 76432-904 76432-906 76432-908 76432-910 76432-912 76432-914 76432-916 76432-918 76432-920 76432-922 76432-924 76432-926 76432-928 76432-930 76432-932 76432-934 76432-936 76432-938 76432-940 76432-942 76432-944 76432-946 76432-948 76432-950 76432-952 76432-954 76432-956 76432-958 76432-960 76432-962 76432-964 76432-966 76432-968 76432-970 76432-972 76432-974 76432-976 76432-978 76432-980 76432-982 76432-984 76432-986 76432-988 76432-990 76432-992 76432-994 76432-996 76432-998 76433-000 76433-002 76433-004 76433-006 76433-008 76433-010 76433-012 76433-014 76433-016 76433-018 76433-020 76433-022 76433-024 76433-026 76433-028 76433-030 76433-032 76433-034 76433-036 76433-038 76433-040 76433-042 76433-044 76433-046 76433-048 76433-050 76433-052 76433-054 76433-056 76433-058 76433-060 76433-062 76433-064 76433-066 76433-068 76433-070 76433-072 76433-074 76433-076 76433-078 76433-080 76433-082 76433-084 76433-086 76433-088 76433-090 76433-092 76433-094 76433-096 76433-098 76433-100 76433-102 76433-104 76433-106 76433-108 76433-110 76433-112 76434-010 76434-012 76434-014 76434-016 76434-018 76434-020 76434-022 76434-024 76434-026 76434-028 76434-030 76434-032 76434-034 76434-036 76434-038 76434-040 76434-042
Avantor® Hichrom HI-5 MS Plus, GC Columns
Chromatography Columns
General purpose GC-MS column with extra stability. For the analysis of solvent impurities, PCBs, hydrocarbons, essential oils, semivolatiles, pesticides, phenols, etc.
Maximum temperature limits may change depending on the film thickness.