Column Type: GC column, Avantor® Hichrom, FAST GC column, Fused silica, Ø int.: 0.10 mm, Length: 10 m, Film Thickness: 0.40 µm
Supplier: VWR International
Column Type: GC column, Avantor® Hichrom, FAST GC column, Fused silica, Ø int.: 0.10 mm, Length: 10 m, Film Thickness: 0.40 µm