Avantor® Hichrom HI-DEX B-03, GC Columns
Supplier: VWR International
76429-854 76429-856 76429-858 76429-860 76429-862 76429-864 76429-866 76429-868 76429-870 76429-872 76430-054 76430-056
76429-854EA 2234.03 USD
76429-854 76429-856 76429-858 76429-860 76429-862 76429-864 76429-866 76429-868 76429-870 76429-872 76430-054 76430-056
Avantor® Hichrom HI-DEX B-03, GC Columns
Chromatography Columns
Unique selectivity for the separation of enantiomers. Useful for the analysis of chiral compounds in fragrances, pesticides and pharmaceuticals in which the determination of the ratio of enantiomers in a sample is necessary.
Max temperature limits may change depending on the film thickness.