Avantor® Hichrom HI-TCEP, GC Columns
Supplier: VWR International
76429-102 76429-104 76429-106 76429-108
76429-102EA 1379.01 USD
76429-102 76429-104 76429-106 76429-108
Avantor® Hichrom HI-TCEP, GC Columns
Chromatography Columns
Application specifc columns ideal for aromatics and oxygenates in gasoline.
Maximum temperature limits may change depending on the film thickness.