Avantor® Hichrom HI-SOLVE 1, GC Columns
Supplier: VWR International
HI62-32-025-50
76429-094EA 353.87 USD
76429-094 76429-096
Avantor® Hichrom HI-SOLVE 1, GC Columns
Chromatography Columns
Application specifc columns developed for complex solvents mixtures analysis. HI-SOLVE 1 is ideal for aromatics and oxygenates in gasoline.
Max temperature limits may change depending on the film thickness.