Column Type: GC column, Avantor® Hichrom, GC column, Fused silica, Ø int.: 0.53 mm, Length: 15 m, Film Thickness: 1.00 µm
Supplier: VWR International
Column Type: GC column, Avantor® Hichrom, GC column, Fused silica, Ø int.: 0.53 mm, Length: 15 m, Film Thickness: 1.00 µm