Avantor® Hichrom HI-35, GC Columns
Supplier: VWR International
76425-612 76425-614 76425-616 76425-618 76425-620 76425-622 76425-624 76425-626 76425-628 76425-630 76425-632 76425-634 76425-636 76425-638 76425-640 76425-642 76425-644 76425-646 76425-648 76425-650 76425-652 76425-654 76425-656 76425-658 76425-660 76425-662 76425-664 76425-666 76425-668 76425-670 76425-672 76425-674 76425-676 76425-678 76425-680 76425-682 76425-684 76425-686 76425-688 76425-690 76425-692 76425-694 76425-696 76425-698 76425-700 76425-702 76425-704 76425-706 76425-708 76425-710 76425-712 76425-714 76425-716 76425-718 76425-720 76425-722 76425-724 76425-726 76425-728 76425-730 76425-732 76425-734 76425-736 76425-738 76425-740 76425-742 76425-744 76425-746 76425-748 76425-750 76425-752 76425-754 76425-756 76425-758 76425-760 76425-762 76425-764 76425-766 76425-768 76425-770 76425-772 76425-774 76425-776 76425-778 76425-780 76425-782 76425-784 76425-786 76425-788 76425-790 76425-792 76425-794 76425-796 76425-798 76425-800 76425-802 76425-804 76425-806 76425-808 76425-810 76425-812 76425-814 76425-816 76425-818 76425-820 76425-822 76425-824 76425-826 76425-828 76425-830 76425-832 76425-834 76425-836 76425-838 76425-840 76425-842 76425-844 76425-846 76425-848 76425-850 76425-852 76425-854 76425-856 76425-858 76430-376 76430-378 76430-380 76430-382 76430-384 76430-386 76430-388 76430-390 76430-392 76430-394 76430-396 76430-398 76430-400 76430-402 76430-404 76430-406 76430-408 76431-140 76431-142 76431-144 76431-146
76425-612EA 896.34 USD
76425-612 76425-614 76425-616 76425-618 76425-620 76425-622 76425-624 76425-626 76425-628 76425-630 76425-632 76425-634 76425-636 76425-638 76425-640 76425-642 76425-644 76425-646 76425-648 76425-650 76425-652 76425-654 76425-656 76425-658 76425-660 76425-662 76425-664 76425-666 76425-668 76425-670 76425-672 76425-674 76425-676 76425-678 76425-680 76425-682 76425-684 76425-686 76425-688 76425-690 76425-692 76425-694 76425-696 76425-698 76425-700 76425-702 76425-704 76425-706 76425-708 76425-710 76425-712 76425-714 76425-716 76425-718 76425-720 76425-722 76425-724 76425-726 76425-728 76425-730 76425-732 76425-734 76425-736 76425-738 76425-740 76425-742 76425-744 76425-746 76425-748 76425-750 76425-752 76425-754 76425-756 76425-758 76425-760 76425-762 76425-764 76425-766 76425-768 76425-770 76425-772 76425-774 76425-776 76425-778 76425-780 76425-782 76425-784 76425-786 76425-788 76425-790 76425-792 76425-794 76425-796 76425-798 76425-800 76425-802 76425-804 76425-806 76425-808 76425-810 76425-812 76425-814 76425-816 76425-818 76425-820 76425-822 76425-824 76425-826 76425-828 76425-830 76425-832 76425-834 76425-836 76425-838 76425-840 76425-842 76425-844 76425-846 76425-848 76425-850 76425-852 76425-854 76425-856 76425-858 76430-376 76430-378 76430-380 76430-382 76430-384 76430-386 76430-388 76430-390 76430-392 76430-394 76430-396 76430-398 76430-400 76430-402 76430-404 76430-406 76430-408 76431-140 76431-142 76431-144 76431-146
Avantor® Hichrom HI-35, GC Columns
Chromatography Columns
Really versatile general purpose mid-polarity phase for a wide range of applications. Ideal in confirmation analysis.
  • Phase composition: Crossbonded Silphenylene - 35% Phenyl - 65% Methyl Polysiloxane
  • Long lifetime
Max temperature limits may change depending on the film thickness.