Reflectance Spectrophotometers, Lovibond®
Supplier: Lovibond
403225 403035 403220 403040
76424-346EA 20860.11 USD
76424-346 76424-348 76424-350 76424-352
Reflectance Spectrophotometers, Lovibond®
Spectrophotometers and Photometers UV/Visible Spectrophotometers
Flexible and robust reflectance spectrophotometer with QA software.
  • Touch screen
  • On-board camera
Model TR 500: An ergonomic, flexible reflectance spectrophotometer with fixed aperture. Touch screen technology with intuitive icons to enable user-specific settings and guide through the measurement process. An on-board camera-based locator and black and white calibration plates ensure accurate measurements and results every time. Multi language support for international use.
Model TR 520: An ergonomic, flexible reflectance spectrophotometer with easily selectable apertures of 10 mm/8 mm and 5 mm/4 mm. Touch screen technology with intuitive icons to enable user-specific settings and guide through the measurement process. An on-board camera-based locator and black and white calibration plates ensure accurate measurements and results every time. Multi language support for international use.
Model TRA 500: An ergonomic, flexible reflectance spectrophotometer with fixed aperture. Touch screen technology with intuitive icons to enable user-specific settings and guide through the measurement process. An on-board camera-based locator and black and white calibration plates ensure accurate measurements and results every time. Multi language support for international use.
Model TRA 520: An ergonomic, flexible reflectance spectrophotometer with easily selectable apertures of 10 mm/8 mm and 5 mm/4 mm. Touch screen technology with intuitive icons to enable user-specific settings and guide through the measurement process. An on-board camera-based locator and black and white calibration plates ensure accurate measurements and results every time. Multi language support for international use.
Designed specifically for the Lovibond® spectrophotometers, the instruments fit tightly into the housing to avoid any ambient lighting entering the sample.