Choice® Cleanroom Face Masks, Berkshire
Supplier: Berkshire
CHFM18040B
76423-872CS 2091.84 USD
76423-872
Choice® Cleanroom Face Masks, Berkshire
Masks Cleanroom Masks
Berkshire masks offer superior filtration efficiency while providing long wearing comfort for operators.
  • Filter medium: 100% polypropylene
  • Non-woven inner layer and outerfacing
  • Fully enclosed comfortable nosepiece
  • Polyester/spandex earloop provides comfort to wearers
  • Polypropylene spunbound, hydrophobic
People are a significant source of contamination in any critical manufacturing process. Choice® cleanroom face masks are designed to protect the cleanroom or controlled environment from human contamination.
Providing excellent particle and bacterial filtration, these 100% polypropylene masks are soft and non-irritating for extended wear. Masks feature a fully enclosed, comfortable nosepiece and polyester/spandex earloops. Ultrasonic bonding reduces contamination and outgassing.
Certifications: Designed for use in ISO class 4-8, Cleanroom class 10 - 100000.
Packaging: Bulk packed; 50 masks per bag.