StickTogether DNA Ligase Buffer, New England Biolabs
Supplier: New England Biolabs (NEB)
B0535S
76423-398EA 50 USD
76423-398
StickTogether DNA Ligase Buffer, New England Biolabs
Buffers Cloning Buffers
StickTogether Buffer is supplied with T3, T7, Hi-T4 and Salt-T4 DNA ligases.
  • At a 1X concentration, this reaction buffer ensures optimal activity of these DNA ligases
  • Provided as a 2X solutionContains 2 vials of buffer
New England Biolabs supplies a reaction buffer with all of its enzymes.