EnzyChrom™ L-Amino Acid Assay Kit, BioAssay Systems
Supplier: BIOASSAY SYSTEMS
ELAA-100
76423-324EA 533.47 USD
76423-324
EnzyChrom™ L-Amino Acid Assay Kit, BioAssay Systems
Assays Nucleic Acid Assays
Quantitative determination of L-amino acids in biological and other samples.
  • Fast and sensitive: linear detection range: 3.3 to 500 µm (colorimetric assay) and 0.13 to 50 µm (fluorimetric assay) for 60 minutes reaction
  • Method: OD 570 nm or FL530/585 nm
  • Samples: serum, culture media, tissue homogenates, cell lysates, urine, food samples, etc
L-amino acids are the building blocks of proteins in biology. Almost all of the common 20 amino acids exist as the L-enantiomer. BioAssay Systems' L-amino acid assay uses an enzyme-catalyzed oxidation of L-amino acids to convert a dye into a colored and fluorescent form. The absorbance at 570 nm or fluorescence intensity at λex/em = 530/585 nm is directly proportional to the L-amino acid concentration in the sample.