Linearity Immunoassay, FLQ, Audit MicroControls, Inc.
Supplier: AUDIT MICROCONTROLS
K947M-5
K948M-5
76423-190EA
491.17
USD
76423-190
76423-192
Linearity Immunoassay, FLQ, Audit MicroControls, Inc.
Controls and Calibration Verification Materials
The Linearity FLQ Fertility for Abbott Systems is an IVD device consisting of ten levels of liquid material and additives in bovine serum albumin. The Linearity FLQ Anemia for Abbott Systems is an IVD device consisting of 5 levels of liquid material and additives in bovine serum albumin.
Each kit has five distinct levels that demonstrate a linear relationship to each other for their respective analytes, reagents and instruments.
Certifications: FDA approved.
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)