HemaPRO Stain Kit, Original; 125 Tests for HP1, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
HP1BSK
76422-822EA 208.28 USD
76422-822
HemaPRO Stain Kit, Original; 125 Tests for HP1, Hardy Diagnostics
Stains Hematology Stains
A Wrights-Giemsa stain, buffer, and rinse for use with the QuickSlide™ HemaPRO automated hematology slide stainer.
  • Produces 125 tests per pack in one self-contained package
The pack contains a special stain formulation for enhanced cell quality.