Everlast™ 247 Rocker, Electron Microscopy Sciences
Supplier: ELECTRON MICROSCOPY SCIENCE
65700-09
76421-792EA 2196.41 USD
76421-792
Everlast™ 247 Rocker, Electron Microscopy Sciences
Shakers and Mixers Rotating/Rocking Shakers
The Everlast™ 247 Rocker is a powerful motor and heavy-duty drive system.
  • Extra large 16×12" work surface
  • Optional stacking platform doubles the working area
  • Platforms made of stainless steel to prevent corrosion or rusting
  • Provides years of uninterrupted service
They are consistent and reliable, even with constant use in cold rooms or incubators. The rocking speed and tilt angle are easily adjustable for gentle or aggressive agitation.