HistoPro® Rapid Tissue Processor, Electron Microscopy Sciences

Supplier: ELECTRON MICROSCOPY SCIENCE
62581-10
76421-772EA 77916.23 USD
76421-772
HistoPro® Rapid Tissue Processor, Electron Microscopy Sciences
Tissue Processors
HistoPro® 600 provides continuous load, rapid tissue processing without microwave artifacts.

  • No microwave artifacts
  • Load up to 100 cassettes every 15 minutes
  • Over 2500 cassettes in an 8-hour shift
  • Compact footprint
  • Built-in fume extraction can connect to lab fume exhaust system
  • Runs on regular power outlet
  • Stir bar and basket agitation

HistoPro® 600 is compact, cost conscious package and easy to use. Processing mode features rotating baskets, continuous load, and programmable station time. Fume control: Activated charcoal filter and connection to external fume hood.

Capacity: 6 Stations; 1 load station; Up to 5 baskets simultaneously (1.8 L reagent capacity)
Display: 20 characters × 8 lines LCD
Programmable: Station times 1 min. to 12 hours
Vacuum: 5 psi (25 cm of Hg) below ambient
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR