Specimen Stages, Electron Microscopy Sciences
Supplier: ELECTRON MICROSCOPY SCIENCE
4500-1 4500-10 4500-11 4500-12 4500-13 4500-14 4500-15 4500-16 4500-17 4500-2 4500-3 4500-4 4500-5 4500-6 4500-7 4500-8 4500-9
76421-736EA 678.34 USD
76421-736 76421-738 76421-740 76421-742 76421-744 76421-746 76421-748 76421-750 76421-752 76421-754 76421-756 76421-758 76421-760 76421-762 76421-764 76421-766 76421-768
Specimen Stages, Electron Microscopy Sciences
Microscope Stages
EMS Specimen Stages that meets most requirements.
These specimen stages are all easy-change, drop-in style with no scews, and are height adjustable, except for the rotary planetary stage.