Orum International TRIO.BAS™ MONO with HEPA Filter, Hardy Diagnostics
Supplier: Hardy Diagnostics
BAS170K BAS171K
76420-906EA 12106.57 USD
76420-906 76420-908
Orum International TRIO.BAS™ MONO with HEPA Filter, Hardy Diagnostics
Samplers Air Samplers
A viable particle air sampling instrument with an attached HEPA filtration system which filters the exhausted air.
  • Features Bluetooth data transfer capability
If your viable air monitoring plan involves transferring the air sampler from different ISO classification environments with varied levels of microbial contamination (ISO Class 7 to ISP Class 5, for instance), a HEPA filter equipped TRIO.BAS™ instrument may be used to avoid transfer of particles from the inside of the aspirating chamber.
Delivery information: Includes TRIO.BAS™ MONO, contact or Petri plate format, Manufacturer's calibration certificate, cable battery charger, stainless steel aspirating head, stainless steel cover head, protection cover for aspirating head; autoclavable, one 10 pk box of HEPA disposable filters, hard shell carrying case to protect the sampler in transit, IQ, OQ, PQ, fillable guideline documents.