Xylene and Chemical Resistant Slide Labels, General Data
Supplier: General Data Healthcare
KXT0524075110
76120-114KT 938.56 USD
76120-114 76120-116 76420-900
Xylene and Chemical Resistant Slide Labels, General Data
Labels
StainerShield XT slide labels are printed using a desktop thermal slide label printer. They produce clear, crisp printed barcodes and text that remain legible, do not fade or smear, and produce successful scans, even after stainer processing.
  • Designed specifically for use in histology labs to positively identify slides
  • Durable labels withstand extended exposure to chemicals and stains
  • Printed text and barcodes do not fade, smear or separate from the slide
They do not require any additional flap or overlay for protection.
.
Delivery information: StainerShield XT Thermal Transfer label kit contains 3 rolls of labels, 1800 labels on each (total 5400 labels), 1 label across the web, wound in on a 1" core. Kit includes one 2.4"×140 m ribbon on a 0.5" core.
Zebra GX430t - High Heat Ribbon kit contains 3 rolls of labels, 1800 labels on each (total 5400 labels), 1 label across the web, wound out on a 1" core. Kit includes two 2.5"×74 m ribbons on a 0.5" core that is 4" wide.