BioClean™ Non Sterile Looped Face Masks
Supplier: Ansell Healthcare
MEA210-0
76420-672CS 776.26 USD
76420-672
BioClean™ Non Sterile Looped Face Masks
Masks Cleanroom Masks
Non sterile, looped face mask with advanced filtration efficiency (bulk-packed).
  • High particle and Bacterial Filtration Efficiency (BFE) provide excellent contimination control
  • Cleanroom compatible materials reduce contamination risks
  • Clip connector for easy and secure fastening
  • Polypropylene/Polyethylene construction
  • Cleanroom class: 10/ISO 4
The BioClean™ Non sterile Looped Face Mask delivers high bacterial, viral, and particle filtration efficiency, while offering greater breathability and comfort for the user. This looped non-sterile face mask can be equipped with a clip connector (clip to purchase separately), which facilitates quick and secure fastening at the back of the head. This cleanroom face mask is processed in a NEBB-certified ISO Class 4 cleanroom environment for reduced contamination risks and personal protection that users can depend on.
Certifications: ASTM F2101; ISO 2859; ISO 9001:2015
Packaging: 100 pieces per sealed inner PE bag; one inner bag per sealed outer PE bag; 20 outer bags per lined carton (2000 pieces).