FDA Approved Digital Oral Thermometer, Propper
Supplier: Propper
26915500
76419-818CS 11.03 USD
76419-818
FDA Approved Digital Oral Thermometer, Propper
Thermometers Digital Thermometers
BT306 digital electronic thermometer is equipped with LCD display with clear reading.
  • Fast and accurate results
  • Easy-to-read temperature in about 60 seconds
  • Last reading memory
  • Fever alarm
  • Auto-shutoff
The accuracy of this thermometer is ±0.2 ºF from 95.9 ºF to 107.6 ºF.