SP Bel-Art H-B DURAC Liquid Crystal Disposable Forehead Stick-On Thermometer Strip, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
B62010-0000
76419-552PK 433.72 USD
76419-552 76419-554
SP Bel-Art H-B DURAC Liquid Crystal Disposable Forehead Stick-On Thermometer Strip, Bel-Art Products, a part of SP
Thermometers Analog Thermometers
Non-invasive body temperature measurement is ideal for workforce personnel.
  • Color-coded readings allow for quick evaluation of human temperature
  • Can be self applied to maintain social distancing during temperature testing
  • Easy to use simply peel off backing and stick to forehead or hand
  • Latex free
  • Utilizes thermocratic liquid crystal technology
  • Disposable
Fast and reliable method to monitor temperature.
Delivery information: Instructions included.