Fetal bovine serum, 50 ml, Premium, United States Origin, Corning®

Supplier: Corning
35-015-CF
76418-024EA 149.38 USD
76418-024
Fetal bovine serum, 50 ml, Premium, United States Origin, Corning®
Sera (Serum)
Fetal bovine serum, United States origin collected at government approved abattoirs in the United States.

  • Shelf life: 60 months
  • Weight: 0.09 kg
  • Format: Liquid

True pooled and triple 0.1 µm sterile filtered that passes sterility, mycoplasma, and 9CFR viral testing requirements.

Granted a Certificate of Suitability, R0-CEP 2018-271, by the European Directorate for the Quality of Medicines (EDQM).
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR