Corning® Transport Medium
Supplier: Corning
Corning transport medium is formulated using the final ingredient concentrations outlined in CDC's SOP#: DSR-052-01 'Preparation of viral transport medium'.
- cGMP grade
- Aspectically sterile-filtered
Transport medium is an isotonic solution at neutral pH containing protective proteins, antibiotic to prevent microbial growth, and anti-mycotic to prevent contamination by yeast and fungi.