Digital Rockers, Thermo Scientific
Supplier: Thermo Fisher Scientific
88882001
76417-528EA 1406.2 USD
76417-528
Digital Rockers, Thermo Scientific
Shakers and Mixers Rotating/Rocking Shakers
Digital Rocker is designed for ELISA, protein synthesis, gel staining and de-staining, antibody staining, and other time-sensitive applications.
  • Digital display of speed and time
  • Platform style: Tray
  • Capacity: 6×96-well plates
  • Timer range: Continuous or 1 s to 9999 minutes
Digital display with big platform, accurate speed control and quick angle adjustment for easy sample agitation. Optional non-slip rubber mat tray is designed for carrying plates, dishes, culture bottles and bags.
Certifications: cCSAus, CE, RoHS.
Ordering information: Two year warranty.
Delivery information: Includes dimpled rubber mat tray, rubber strip, user manual, EU plug, UK plug, CHN plug, US/Japan plug.