Calibration weights, OIML M1
Supplier: Mettler Toledo
30406445 30406446 30406447 30406448 30406449 30406450 30406451 30402721 30402715 30402732 30402682 30402685 30402679 30402728
76417-420EA 369 USD
76417-420 76417-422 76417-424 76417-426 76417-428 76417-430 76417-432 76417-496 76417-498 76417-500 76417-502 76417-504 76417-506 76417-508
Calibration weights, OIML M1
Weights Calibration Weights
Set M1 OIML class weight in a plastic box including accessories for handling and cleaning and a calibration certificate.
  • Stainless steel weights
Each knob weight has an adjusting cavity, which simplifies weight adjustment without the need for specialized equipment.