OIML Class M1 Standard Line Weights
Supplier: Mettler Toledo
30402184 30402510 30402355 30402585 30402586 30402587 30402638 30402639 30402640 30402641 30402642 30402643 30402644 30402645 30402646 30402647 30402576 30402699 30402702 30402665 30402708 30402711 30402780
76417-442EA 20.15 USD
76417-442 76417-444 76417-446 76417-448 76417-450 76417-452 76417-454 76417-456 76417-458 76417-460 76417-462 76417-464 76417-466 76417-468 76417-470 76417-472 76417-474 76417-476 76417-478 76417-480 76417-482 76417-484 76417-486
OIML Class M1 Standard Line Weights
Weights Calibration Weights
Single M1 OIML class sheet and knob weights in a plastic box for standard environments.
  • Affordable quality weights for regular routine testing without the need for traceability
  • No calibration certificate
  • 304 Stainless steel
  • <0.8 Susceptibility X
  • 7950 (±140) kg/m³ density ρ
Adjusting cavity: Each knob weight has an adjusting cavity, which simplifies weight adjustment without the need for specialized equipment.