OIML Class M1 Advanced Line Weights
Supplier: Mettler Toledo
30406452 30406453 30406454 30406455 30406456 30406457 30406458 30406459 30406460 30406371 30406372 30406373 30406374 30406375 30406376 30406377 30406478 30406479 30406480 30406481 30406482 30406483 30406484
76417-374EA 59.29 USD
76417-374 76417-376 76417-378 76417-380 76417-382 76417-384 76417-386 76417-388 76417-390 76417-392 76417-394 76417-396 76417-398 76417-400 76417-402 76417-404 76417-406 76417-408 76417-410 76417-412 76417-414 76417-416 76417-418
OIML Class M1 Advanced Line Weights
Weights Calibration Weights
Single M1 OIML class sheet and knob weight in a plastic box including a calibration certificate.
  • Material: 304 Stainless steel
  • Density ρ: 7950 (±140) kg/m³
  • Susceptibility X: <0.8
Our investments in state-of-the-art robot technology, combined with our many years of experience, minimize errors and increase accuracy.
Each knob weight has an adjusting cavity, which simplifies weight adjustment without the need for specialized equipment.
Certifications: All weights are delivered with a Calibration Certificate accordance with ISO/IEC 17025 requirements.