VWR® Screw Cap Microtubes
Supplier: VWR International
76417-204 76417-206 76417-208 76417-210 76417-212 76417-214 76417-232 76417-234 76417-236 76417-238 76417-240 76417-242 76417-244 76417-246 76417-248 76417-250 76417-252 76417-254 76417-256 76417-258
76417-204CS 2078.49 USD
76417-204 76417-206 76417-208 76417-210 76417-212 76417-214 76417-232 76417-234 76417-236 76417-238 76417-240 76417-242 76417-244 76417-246 76417-248 76417-250 76417-252 76417-254 76417-256 76417-258
VWR® Screw Cap Microtubes
Tubes Microtubes
VWR® screw cap microtubes are the perfect environment to securely store your important samples and reagents.
  • All tubes are manufactured from ultra-pure PP
  • High-clarity tubes provide excellent visibility to the sample
  • O-ringed flat caps feature a ³/₄ turn to open or close
  • Wide temperature range to meet a wide variety of applications: −196 to +121 °C, but not suitable for immersion in liquid nitrogen
  • Sterile assemblies or handy, non sterile combo packs are available
Screw cap microtubes are available in the three most commonly volumes and available with conical or free-standing/skirted bottoms.
Certifications: Certified free of detectable DNase, RNase, DNA, PCR inhibitors. Sterile items are also free of pyrogens. The PP used in tubes meets FDA 21 CFR 177.1520 & USP Class VI standards.