VWR® Screw Cap Microtubes with Customizable Caps
Supplier: VWR International
76417-160 76417-162 76417-164 76417-166 76417-168 76417-170 76417-172 76417-174 76417-176 76417-178 76417-180 76417-182
76417-160CS 1756.7 USD
76417-160 76417-162 76417-164 76417-166 76417-168 76417-170 76417-172 76417-174 76417-176 76417-178 76417-180 76417-182
VWR® Screw Cap Microtubes with Customizable Caps
Tubes Microtubes
VWR® screw cap microtubes are the perfect environment to securely store your important samples and reagents.
  • Manufactured from ultra-pure PP
  • High-clarity tubes offer excellent visibility to the sample
  • Wide temperature range to meet a wide variety of applications: –196 to +121 °C, but not suitable for immersion in liquid nitrogen
  • Available in sterile assemblies or handy, non sterile combo packs
  • Customizable caps accept colorful cap inserts to organize samples or match kit branding
VWR® screw cap microtubes are available in the three most commonly volumes and available with conical or free-standing/skirted bottoms.
Certifications: Certified free of detectable DNase, RNase, DNA, PCR inhibitors. Sterile items are also free of pyrogens. The PP used in tubes meets FDA 21 CFR 177.1520 & USP Class VI standards.
Ordering information: Spare caps and color insert chips also available.