CiDecon® II Concentrated Phenolic Liquid Disinfectant, Decon Labs
Supplier: Decon Labs
8514
76417-136CS 511.45 USD
76417-136
CiDecon® II Concentrated Phenolic Liquid Disinfectant, Decon Labs
Cleaning Agents Disinfectants
CiDecon® II phenolic-based disinfectant is a concentrated liquid packaged in 1 gallon bottles.
  • Phenolic-based concentrated disinfecant
  • Dilution is 1 ounce per gallon of water (1:128)
CiDecon® II concentrated phenolic disinfectant is a high pH (alkaline) germicidal cleaner for use on hard surfaces in labs, production areas, on process equipment, or anywhere thorough disinfection efficacy is required.
CiDecon® II can be used for biological decontamination and disinfection. It is effective against a wide range of pathogenic organisms including TB, HIV-1, bacteria, viruses, and fungi. High soil-load and hard water tolerance. It can be alternated with LopHene II if a rotational disinfection program is desired, or where organism resistance is suspected. Conforms to OSHA’s blood borne pathogens standard. EPA registered.