Lead/Latex-Free Steam Indicator Tape, 7158, UltraTape
Supplier: Ultratape Industries
7158ST100-P6S
76416-362CS 675.87 USD
76416-362
Lead/Latex-Free Steam Indicator Tape, 7158, UltraTape
Sterilization Indicators
The 7158 steam indicator tape is a latex-free, lead-free high tensile, premium grade, saturated crepe paper with a tinted stripe of color changing ink that produces a dramatic color change upon autoclave process completion.
  • Saturated crepe paper backing
  • Synthetic solvent-free adhesive
  • Natural finish with green printed lines
  • Latex-free and lead-free
  • Operating temperature: 120 °C for 30 min, 132 °C for 4 min and 135 °C for 3 min
Autoclavable steam indicator paper is designed for high temperatures. It will adhere to non-woven and muslin fabric as well as plastic and metals.