AMI Illuminated Magnetic Stirrers, Velp Scientific
Supplier: VELP SCIENTIFIC INC.
F204A0167
76416-230EA 674.78 USD
76416-230
AMI Illuminated Magnetic Stirrers, Velp Scientific
Stirrers Magnetic Stirrers
Single-position illuminated magnetic stirrer, particularly useful for titrations.
  • Control panel is shielded from liquid spills
  • Epoxy-coated tough steel casing
  • Soft start but rapidly reaching set speed
  • Excellent speed control (electronic), even at low speed
  • Possibility to adjust the illumination brightness
  • Ergonomic - very low height
  • Universal mains voltage range, light weight
Illuminated surface for visualization of particles and color changes. Ideal for microtitration, in particular for subtle color changes. Four-position model provides independent speed regulation. High-power driving magnet type 'PCM' operated by a mono-phase motor for continuous operation stirring system.