MST Magnetic Stirrer, Velp Scientific
Supplier: VELP SCIENTIFIC INC.
F203A0440
76416-218EA 151.08 USD
76416-218
MST Magnetic Stirrer, Velp Scientific
Stirrers Magnetic Stirrers
Small and efficient magnetic stirrer.
  • Technopolymer casing resists chemicals
  • White surface for visualization of particles and color changes
  • Speed regulation up to 1100 rpm
  • Accepts up to 5 L flasks
  • Remains cold, even after days of continuous use
  • Ideal for microtitration, BOD, microbiology and biochemistry
  • Excellent high power magnet type PCM speed control, even at low speeds
Delivery information: Three year warranty.