Vortex Mixer, CLASSIC, VELP Scientifica
Supplier: VELP SCIENTIFIC INC.
F202A0173
76416-198EA 392.65 USD
76416-198
Vortex Mixer, CLASSIC, VELP Scientifica
Shakers and Mixers Vortex Mixers
Vortex mixer suitable for different mixing requirements, with touch or continuous mode.
  • Robust and convenient
  • Compact in height and area
  • Stability is optimized with selection lever related to operating mode
  • Three anti-slip rubber feet for staying in required position
  • Variable speed operation (electronically controlled)
Universal voltage or frequency model. Optimum resistance to chemicals from technopolymer casing. Excellent stability with zinc alloy base. Orbital movement with analog speed setting.
Certifications: IP Protection Class according toMore Product Information