Portable Hand Sink with Open Base, Eagle MHC
Supplier: Eagle Group
PHS-E-S-H
76416-054EA 6061.03 USD
76416-054
Portable Hand Sink with Open Base, Eagle MHC
Faucets and Sinks Sinks
Portable hand sinks need no additional plumbing, all you need is water and a 120-volt receptacle
  • Stainless steel tabletop and stainless steel undershelf
  • 5-gallon fresh water tank and 7-gallon waste water tank
  • Pump with 150 V, GFCI electrical outlet and 10' cord/plug with cord wrap
  • Deck mount faucet, soap dispenser and towel dispenser
  • Removable pan, 10⅜×12¾×6"
This portable hand sink offers a mobile solution for adding handwashing stations in unconventional locations, or where they are needed on a temporary basis. Everything is self-contained on a portable hand sink.