CompactDry™ Yeast/Mold (YM), Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
CompactDry™ YM is a ready-to-use test method recommended for the determination of total aerobic bacterial counts.
- Sturdy frame, stackable packaging
- Self-diffusing media
- Ability to subculture without damage to colonies
- No leakage
- Easy to label
CompactDry™ is a ready-to-use system that reduces the time needed to perform microbial testing on food, cannabis, meat, cosmetics, raw materials and more.
Each cassette comes with 4 connected plates that can be easily snapped apart to support a variety of dilution scenarios.
Caution: Stored at room temperature vs the competitor products with storage at temperatures less than or equal to 8 °C.