CompactDry™ Total Count (TC), Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
54081
76414-994EA
173.04
USD
76414-994
CompactDry™ Total Count (TC), Hardy Diagnostics
Microbiology Test Systems
Microorganism Tests
CompactDry™ TC is a ready-to-use test method recommended for the determination of total aerobic bacterial counts.
- No leakage
- Easy to label
- Self-diffusing media
- Sturdy frame, stackable packaging
- Ability to subculture without damage to colonies
Each cassette comes with 4 connected plates that can be easily snapped apart to support a variety of dilution scenarios.
Caution: Stored at room temperature vs the competitor products with storage at temperatures less than or equal to 8 °C.