CompactDry™ Salmonella (SL), Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
54085
76414-992EA 262.1 USD
76414-992
CompactDry™ Salmonella (SL), Hardy Diagnostics
Microbiology Test Systems Microorganism Tests
CompactDry™ SL is a ready-to-use test method recommended for the determination of total aerobic bacterial counts.
  • Sturdy frame, stackable packaging
  • Self-diffusing media
  • Ability to subculture without damage to colonies
  • No leakage
  • Easy to label
Each cassette comes with 4 connected plates that can be easily snapped apart to support a variety of dilution scenarios.
Caution: Stored at room temperature vs the competitor products with storage at temperatures less than or equal to 8 °C.