Defender II Disposable Face Shield, Biotix
Supplier: Biotix
63305282
76414-911CS 180.36 USD
76414-911
Defender II Disposable Face Shield, Biotix
Face Shields
The Defender II face shield is intended to protect healthcare providers who are in close contact with humans, samples (aerosols, biofluids, biosolids), and other biohazardous materials or dangerous chemicals.
  • Ergonomic design, with a lightweight and comfortable forehead pad
  • Protects against airborn pathogens, liquid/solid samples
  • Offers distortion-free visibility (central and peripheral vision)
  • Compatible with other PPE (goggles, safety glasses, mask)
  • Anti-fog design, with open sides and anti-fog coating
  • Easy to clean
  • Latex-free, anti-static, and nonsterile
The face shield combines exceptional comfort (lightweight, anti-fog design and coating, and foam pad for comfort and airflow) and safety (latex-free, protects face and neck). The elastic headband ensures an easy fit no matter the size of the head and face. The headgear also features a forehead sponge for increased comfort during extended use.
Certifications: OSHA compliant to ANSI 87.1:2015.