Thunder Imager Assay Packages, Leica Microsystems
Supplier: Avantor
76414-906EA 168534.09 USD
76414-906
Thunder Imager Assay Packages, Leica Microsystems
Microscopes
THUNDER imager assay is an innovative solution for advanced 3D cell culture assays, whether for analyzing stem cells, spheroids, or organoids.
Unique automation features accelerate imaging of large sample volumes for more accurate statistics and minimizes user interaction (even for complex experiments) to improve assay workflow efficiency. High sensitivity of the system ensures low phototoxicity and photobleaching, resulting in higher data throughput with optimal physiological conditions for your sample. With Leica developed computational clearing, THUNDER efficiently removes out-of-focus blur in real time, enabling the meaningful use of 3D specimens with camera-based widefield fluorescence microscopes.
Take 3D cell culture research to the next level with groundbreaking ease-of-use, speed and sensitivity with THUNDER Imager Assay.
Certifications: CE and CAS.