THUNDER Imager Tissue Packages, Leica Microsystems
Supplier: Avantor
76414-904EA 70422.75 USD
76414-904
THUNDER Imager Tissue Packages, Leica Microsystems
Microscopes
The THUNDER imager tissue allows real time fluorescence imaging of 3D tissue sections typically used in neuroscience and histology research. Acquire rich, detailed images of thick tissues free of haze from ‘out of focus’ blur.
Certifications: CE and CAS.
Delivery information: THUNDER imager tissue imager package includes: DM4 B semi-motorised upright microscope stand, manual camera port, motorised 5-position fluorescence filter turret, coded 7-position objective nosepiece, Objectives (Plan FL 1.25x/0.04NA, Plan FL 10x/0.32NA and Plan Apo 40x/0.95NA with correction collar), 10x/25 mm FOV eyepieces, LED transmitted light, Brightfield condenser (0.90 NA), Manual stage (110° rotation), shutter controlled LED3 fluorescence light source, Fluorescence filters (LED405, GFP, and RHOD), K5 sCMOS camera, Dust cover, THUNDER Workstation 2D, US Keyboard, LAS X Core software, LAS X Multi-Channel Acquisition software module, 32" monitor and 1 TB SSD RAID addition.
Purchase of THUNDER Imager Tissue Package includes service installation and 1 day of applications training by LMS Advanced Workflow Specialist.