Azure Aqua Transfer Cell
Supplier: Azure Biosystems
AC4201
76414-792EA 684.94 USD
76414-792
Azure Aqua Transfer Cell
Electrophoresis Reagents Western Blotting Reagents
The Azure Aqua transfer cell is used for transferring two mini gels to membranes for Western blotting experiments (wet transfer).
  • W×D×H: 10×18×10 cm
The transfer cassettes and electrode core are colored for directionality which makes transferring easy and straightforward. You can perform a quick one hour transfer or you can transfer overnight at a lower voltage.
Ordering information: Transfer cell includes 2 gel transfer cassette holders, 6 sponges, 2 Azure Aqua cooling units, electrode assembly, tank and top lid with power cables.