Filtering Bottle, United Scientific Supplies
Supplier: United Scientific Supplies
FFB5340-20000
76414-742EA 790.06 USD
76414-742
Filtering Bottle, United Scientific Supplies
Flasks Filter Flasks
High-quality large volume filtering flask.
  • Graduated in 2000 ml increments on a scale from 4000 to 16000 ml with an accuracy of ±5%
  • Neck size fits a #13 rubber stopper
  • Hose barb outer Ø of ¹/₂"
With its thick glass walls and sturdy construction, this 20 L bottle/flask is perfect for filtering large volumes of liquid. The thick walls and design enables this flask to withstand vacuum filtration without imploding.