Low Form Beakers, United Scientific Supplies
Supplier: United Scientific Supplies
BG1000-10000 BG1000-20000
76414-728EA 401.29 USD
76414-728 76414-730
Low Form Beakers, United Scientific Supplies
Beakers
Quality beaker made from low-expansion borosilicate glass.
  • Heavy and uniform wall thickness
  • Spout for easy pouring
  • Beaded tops
The 20 ml and larger sizes also have a double graduated metric scale and a marking area. Tolerance for beakers is approximately ±5%.