Test Tube Rack, Marlin Steel Wire Products
Supplier: Marlin Steel Wire Products
00689055-39
76414-152EA 288.29 USD
76414-152
Test Tube Rack, Marlin Steel Wire Products
Racks Tube Racks
Quality engineered test tube racks used for sterilzation, cleaning, and research.
  • Holds 72 test tubes that are 18 mm (0.709") diameter tubes
  • Stainless Steel 316 grade with natural finish
Typically used for autoclaving test tubes.