Dispersing Aggregates for POLYTRON® System PT10-35 GT, Kinematica
Supplier: Kinematica
11030305
76414-144EA 4121.98 USD
76414-144
Dispersing Aggregates for POLYTRON® System PT10-35 GT, Kinematica
Homogenizers Dispersing Tools
Dispersing aggregates available as standard (EC design) and special aggregates (W, B design).
  • Manufactured from high-alloy stainless steel 316L, electrolytically polished
  • Simple removal and fast cleaning either by hand, dishwasher or autoclave
  • Easy disassembly of components
  • Fitted with a high-precision quick-release system
Standard aggregates: Useful in the preparation of samples and formulation development. The cutting teeth embedded in the head of the tool optimise the pre-crushing of the sample. An enclosed ring design provides optimum protection for those teeth and prevents them from bending.
Special aggregates: Especially for use in the chemical, pharmaceutical, cosmetic, food industry and life sciences.