NANOSTAR 7.5 Digital Electronic Overhead Stirrer, IKA
Supplier: IKA Works
0025004356
76414-098EA 1796.96 USD
76414-098
NANOSTAR 7.5 Digital Electronic Overhead Stirrer, IKA
Stirrers Overhead Stirrers
Small and space saving overhead stirrer with great performance.
  • Guarantees a constant torque over the entire rpm range from 50 to 2000 min⁻¹
  • Easy to operate using a stable rotary knob
  • Display clearly shows the rotational speed
  • Regular software updates can be undertaken rapidly and simply via the USB interface
The NANOSTAR 7.5 digital overhead stirrer is 30% smaller than the MICROSTAR 7.5 digital and therefore even more space-saving. However, its performance remains the same. NANOSTAR 7.5 digital convinces with perfect basic functionalities, its compact design and weighing just 0.8 kg means it is easy to handle.
Certifications: IP protection class according to DIN EN 60529: IP 54.
Ordering information: Supplied without accessories, a wide range of accessories are available to order separately.