AlphaTec® Neoprene Isolator Gloves, 55-305 Series, Ansell
Supplier: Ansell Healthcare
55305100
76413-634CS 3590.09 USD
76413-634
AlphaTec® Neoprene Isolator Gloves, 55-305 Series, Ansell
Gloves
Non-clean/non sterile neoprene RABS/Isolator gloves for use in 254 mm (10") ports.
  • Effective protection for medium duty chemical environments
  • Port size: 254 mm (10")
  • Exceptional chemical resistance against most acids, alcohols, oils, lubricants, and hydrocarbons
  • Medium thickness (20 mil/0.51 mm) increases dexterity, which improves the ability to manipulate objects
  • Unlined for effective cleaning
AlphaTec® Neoprene Isolator Gloves 55-305 are EN ISO 374-certified for chemical resistance, offering wearer protection against most acids, alcohols, oils, and lubricants. The unlined design of AlphaTec®, Neoprene Isolator Gloves 55-305 lessens the chance of chemical absorption and related contamination risks. These neoprene gloves also feature a practical, ambidextrous design, making donning easy. The 55-305 glove series features EN ISO 388 certification for abrasion resistance, granting them greater durability.
Certifications: CE 0493, EN 374:2003, EN 388:2003, EN 388:2016 +A1:2018, EN ISO 21420:2020, EN ISO 374-1:2016, EN ISO 374-5:2016, EN ISO 374-1:2016, EN ISO 374-5:2016, EN 388:2016, EN 420:2003 + A1:2009, Category III, UKCA 0321.
Packaging: 1 pair per black sealed plastic bag; 10 pairs per shipper carton.