AlphaTec® Neoprene Isolator Gloves, 55-303 Series, Ansell
Supplier: Ansell Healthcare
55303100
76413-632CS 2314.55 USD
76413-632
AlphaTec® Neoprene Isolator Gloves, 55-303 Series, Ansell
Gloves
Non-clean/non sterile neoprene RABS/Isolator gloves for use in 203 mm (8") ports.
  • Effective protection for heavy duty chemical environments
  • Port size: 203 mm (8")
  • Exceptional chemical resistance against most acids, alcohols, oils, lubricants and hydrocarbons
  • Heavyweight thickness (30 mil/0.76 mm) increases durability, augmenting chemical and physical resistance
  • Unlined for effective cleaning
AlphaTec® Neoprene RABS and Isolator gloves deliver exceptional chemical resistance against most acids, alcohols, oils, lubricants, and hydrocarbons. They offer strong physical protection against abrasions and good protection against cuts and punctures.
Certifications: CE 0493, EN 374:2003, EN 388:2003, EN 388:2016 +A1:2018, EN ISO 21420:2020, EN ISO 21420:2020, EN ISO 374-1:2016, EN ISO 374-5:2016, EN ISO 374-1:2016, EN ISO 374-5:2016, EN 388:2016, EN 420:2003 + A1:2009, Category III, UKCA 0321.
Packaging: 1 pair per black sealed plastic bag; 10 pairs per shipper carton.